missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine-Bromine Solution, Hanus, for Iodine Absorption Number of Fats and Oils, Ricca Chemical
For Analysis of Iodine Absorption Number of Fats and Oils
$177.03 - $692.50
Chemical Identifiers
| CAS | 64-19-7 |
|---|---|
| Molecular Formula | C2H4O2 |
| Molecular Weight (g/mol) | 60.05 |
| MDL Number | MFCD00036152 |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
| PubChem CID | 176 |
| ChEBI | CHEBI:15366 |
| IUPAC Name | acetic acid |
| SMILES | CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
4100-4
|
Ricca Chemical Company
RI41004 |
120 mL | Amber Glass Bottle |
N/A
|
|
||||
|
4100-16
|
Ricca Chemical Company
410016 |
500 mL | Amber Glass Bottle |
N/A
|
|
||||
|
4100-32
|
Ricca Chemical Company
410032 |
1 L | Amber Glass Bottle |
N/A
|
|
||||
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Specifications
| Brown | |
| 96.0,1.21,0.85 | |
| C2H4O2 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| Amber Glass Bottle | |
| 120 mL | |
| Iodine-Bromine Solution, Hanus |
| 64-19-7,7553-56-2,7726-95-6 | |
| 100.0,1.26,0.89 | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| Laboratory | |
| <1 | |
| Liquid |