missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine-Bromine Solution, Hanus, for Iodine Absorption Number of Fats and Oils, Ricca Chemical
Specifications
Specifications
| Color | Brown |
| CAS Min % | 96.0, 1.21, 0.85 |
| CAS Max % | 100.0, 1.26, 0.89 |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| pH | <1 |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Chemical Name or Material | Iodine-Bromine Solution, Hanus |