missing translation for 'onlineSavingsMsg'
Learn More

Iodine-Bromine Solution, Hanus, for Iodine Absorption Number of Fats and Oils, Ricca Chemical

For Analysis of Iodine Absorption Number of Fats and Oils

Supplier:  Ricca Chemical Company 410016

 View more versions of this product

Catalog No. 4100-16


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Iodine-Bromine Solution
Hanus
96.0,1.21,0.85
C2H4O2
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
60.05
CHEBI:15366
Amber Glass Bottle
500 mL
Brown
64-19-7,7553-56-2,7726-95-6
100.0,1.26,0.89
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
Laboratory
<1
Liquid

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions