missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 0.100 Normal (N/10), Reagents

Numéro de catalogue. 01336482
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01336482 1 L
01336481 2.5 L
01336480 4 L
01336479 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 01336482 Fournisseur Labchem Inc Code fournisseur C72176001C
Il en reste null

Iodine, 0.100 Normal (N/10), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Identifiants chimiques

CAS 7732-18-5 , 7681-11-0 , 7681-11-0
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Diiodine solution
IUPAC Name water
SMILES O

Spécifications

Boiling Point 100°C
Color Dark Brown
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.05
Chemical Name or Material Iodine, 0.100 Normal (N/10)