missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iodine, 0.01N Standardized Solution

CAS: 7553-56-2 | I2 | 253.81 g/mol

Supplier:  Thermo Scientific Chemicals 035636K2

 View more versions of this product

Catalog No. AA35636K2


Only null left
Explore available promotions

Description

Description

Iodine, 0.01N standardized solution is used for accurate volumetric analysis. It is also used as a disinfectant and antibacterial agent in solutions, tinctures and salves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iodine
23 hPa (17mm Hg) at 20°C
I2
Red to Brown
PNDPGZBMCMUPRI-UHFFFAOYSA-N
diiodine
1 L
CHEBI:17606
Iodine: 0.13%; Potassium iodide: 2.5%; Water: 97.37%
7553-56-2 , 7681-11-0 , 7732-18-5
Liquid
MFCD00011355 MFCD00164163
Miscible with chloroform, carbon tetrachloride, carbon disulfide, cyclohexane, methanol, ethyl acetate, toluene, benzene, n-hexane, butan-2-ol, bromoethane, n-heptane, glycerol and diethyl ether
II
253.81
807
253.81
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only