missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iodine, 0.01N Standardized Solution

Catalog No. p-4408552
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35636K2 1 L
AA35636K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AA35636K2 Supplier Thermo Scientific Chemicals Supplier No. 035636K2
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 7553-56-2 | I2 | 253.81 g/mol

Iodine, 0.01N standardized solution is used for accurate volumetric analysis. It is also used as a disinfectant and antibacterial agent in solutions, tinctures and salves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Iodine
CAS 7553-56-2, 7681-11-0, 7732-18-5
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
Color Red to Brown
Solubility Miscible with chloroform, carbon tetrachloride, carbon disulfide, cyclohexane, methanol, ethyl acetate, toluene, benzene, n-hexane, butan-2-ol, bromoethane, n-heptane, glycerol and diethyl ether
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name diiodine
Molecular Weight (g/mol) 253.81
Quantity 1 L
PubChem CID 807
ChEBI CHEBI:17606
Formula Weight 253.81
Concentration or Composition (by Analyte or Components) Iodine: 0.13%; Potassium iodide: 2.5%; Water: 97.37%
Show More Show Less

RUO – Research Use Only