missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RadPrime DNA Labeling System

Catalog No. 18428011
Encompass
Change view
Click to view available options
Quantity:
30 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18428011 30 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18428011 Supplier Invitrogen™ Supplier No. 18428011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for rapid preparation (under 10 min) of high-specific activity 32 P-labeled probes for the detection of DNA and RNA

  • Produces probes that can detect nucleic acids on northern or Southern blots, plaque lifts, colony hybridizations, and in situ hybridization
  • Yields >109cpm/μg control DNA using [32P]-dCTP
  • Labels 25ng of DNA in one reaction
  • Performance and Quality Testing: Incorporation of a radioactively labeled nucleotide is verified using control DNA in a RadPrime labeling reaction

DNA and RNA Purification and Analysis, DNA Labeling, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Nucleic Acid Labeling and Oligo Synthesis, Southern Blotting

Order Info

Shipping Condition: Dry Ice

Specifications

Product Type DNA Labeling System
Detection Method Radioactive
Content And Storage Components of the RadPrime DNA Labeling System are listed in the table to the right. Store at -20°C.
.
Final Product Type Probes (Labeled DNA)
Label or Dye 32P (Phosphorous-32)
Includes Label or Dye No
Labeling Target DNA (General)
Labeling Method Direct Labeling
Shipping Condition Dry Ice
Quantity 30 Reactions
How long does the labeling take when using the RadPrime DNA Labeling system?

Typically labeling is complete in 10 minutes, and purifiation of the probe is not usually required.

For Research Use Only. Not for use in diagnostic procedures.