missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Proteinase K Solution (20 mg/mL), RNA grade

Catalog No. 25530049
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25530049 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25530049 Supplier Invitrogen™ Supplier No. 25530049

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.

  • Over a wide pH range—optimal activity between 6.5 and 9.5
  • Under denaturing conditions—e.g., in the presence of SDS or urea
  • In the presence of metal chelating agents—e.g., EDTA
  • At comparatively high temperatures—optimum digestion temperature is 65°C
  • Protein Filter: P

Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and quality testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit definition
One mAnson unit is described as that amount of enzyme that liberates 1 μmol of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

Specifications

Concentration 20 mg/mL
Purity DNase-Free, RNase-Free
For Use With (Application) Chromatin Biology/RNA Extraction/DNA Extraction
Content And Storage Store in freezer (-5 to -30°C).
Shipping Condition Approved for shipment on Wet or Dry Ice
Quantity 5 mL
Product Type Proteinase K
Form Liquid
How do you recommend storing proteinase K?

We recommend storing it at 4°C.

How do I isolate DNA from single bacterial colonies before PCR?

Scrape a single colony from the surface of an agar plate and transfer to 12 µL of SCL solution [10 mM Tris-HCl (pH 7.5), 1 mM EDTA, 50 µg/mL proteinase K] and incubate for 15 min at 55 degrees C. Inactivate proteinase K for 15 min at 80 degrees C and add 20 µL of deionized water. Centrifuge at 12,000 x g for 3 min. Transfer the supernatant to a new tube.

What are the recommended conditions for proteinase K treatment when isolating RNA or DNA samples?

Concentration: Generally proteinase K is used in the concentration range of 50 to 500 µg/mL at 65 degrees C in the presence of SDS (0.5-1%).

Temperature optimum: 65 degrees C; 12X more active at 65 degrees C than at 25 degrees C.

pH: Proteinase K is stable over a wide pH range (4.0 to 12.5), with optimal activity at pH 6.5 to 9.5. It is most stable at pH 8.
Inactivation: Heat inactivate Proteinase K at 80 degrees C for 15 min. Phenol extraction is the recommended method to ensure complete inactivation.

Inhibited by: PMSF (0.1 to 1.0 mM of PMSF is usually sufficient for inhibition of Proteinase K)

Not inhibited by: Proteinase K is not inactivated by metal ions, chelating agents (e.g., EDTA), sulfhydryl reagents or by trypsin or chymotrypsin inhibitors. Activity can be stimulated by addition of denaturing agents (SDS and urea).

For Research Use Only. Not for use in diagnostic procedures.