missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ TOP10F' Chemically Competent E. coli

Catalog No. C303003
Encompass
Change view
Click to view available options
Quantity:
21 x 50 μL
42 x 50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C303003 21 x 50 μL
C303006 42 x 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C303003 Supplier Invitrogen™ Supplier No. C303003

One Shot TOP10F' Chemically Competent E. coli are identical to TOP10 cells, with the addition of an F' episome.

One Shot TOP10F´ Chemically Competent E. coli are identical to TOP10 cells, with the addition of an F´ episome. The F´ episome carries a tetracycline resistance gene and allows isolation of single-stranded DNA (ssDNA) from vectors that have an f1 origin of replication. In addition, the F´ episome carries the lacIq repressor for inducible expression from trc, tac, and lac promoters using IPTG.

TOP10F´ competent cells are provided at a transformation efficiency of >1 x 109 cfu/μg supercoiled DNA and are ideal for high-efficiency cloning and plasmid propagation.

As a derivative of TOP10, TOP10F´ Chemically Competent E. coli cells carry mutations in the methylation-dependent restriction system (mcrA, mcrBC, and mrr) allowing them to clone both prokaryotic and eukaryotic genomic DNA and provide efficient plasmid rescue from eukaryotic genomes. TOP10F´ cells also carry recA1 and endA1 mutations that increase insert stability and improve the quality of plasmid DNA prepared from minipreps.

One Shot TOP10F´ Chemically Competent E. coli cells offer:
• Transformation efficiencies of >1 x 109 cfu/μg
• Stable F´ episome that allows isolation of ssDNA and carries lacIq repressor for inducible expression from trc, tac, and lac promoters
hsdR for efficient transformation of unmethylated DNA from PCR amplifications
mcrA for efficient transformation of methylated DNA from genomic preparations
lacZΔM15 for blue/white color screening of recombinant clones on plates containing X-Gal or Bluo-Gal
endA1 for cleaner DNA preparations and better results in downstream applications due to elimination of nonspecific digestion by Endonuclease I
recA1 for reduced occurrence of nonspecific recombination in cloned DNA

Note: IPTG induction is required for blue-white color screening.

Easy-to-use One Shot format
TOP10 F´ Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format allows all steps of the transformation protocol, up to plating, to take place in the same tube, saving time and preventing contamination.

Genotype
F´{lacIq, Tn10(TetR)} mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK rpsL(StrR) endA1 nupG

Find the strains and formats that fit your needs
• DH strains are available in chemically competent and electrocompetent cell formats.
• For high throughput applications there are a variety of TOP10 strains in MultiShot formats.
• For ssDNA production try strains such as TOP10F´ (Cat. No. C303003) or electrocompetent DH12S (Cat. No. 18312017).

Order Info

Shipping Condition: Dry Ice

Specifications

Product Type Chemically Competent Cells
Contains F' Episome Yes
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • One Shot TOP10F' Chemically Competent E. coli (21 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 plasmid (50 μL at 10 pg/μL)
Store pUC19 plasmid at –20°C.

• S.O.C. Medium (6 mL)
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial Yes (Streptamycin, Tetracycline)
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Promoter Trc, Tac, Lac
Format Tube
Product Line One Shot
Quantity 21 x 50 μL
Show More Show Less
What is the difference between TOP10 and TOP10F' cells?

The only difference between TOP10 and TOP10F' cells is that the latter contain the F' episome that carries the tetracycline resistance gene and allows isolation of single-stranded DNA from vectors that have an f1 origin of replication. The F' episome also carries the lacIq repressor for inducible expression from trc, tac, and lac promoters using IPTG. TOP10F' cells require IPTG induction for blue/white screening.

I am trying to clone an insert that is supposedly pretty toxic. I used DH5? and TOP10 cells for the transformation and got no colonies on the plate. Do you have any suggestions for me?

If the insert is potentially toxic to the host cells, here are some suggestions that you can try:

- After transforming TOP10 or DH5? cells, incubate at 25-30°C instead of 37°C. This will slow down the growth and will increase the chances of cloning a potentially toxic insert.
- Try using TOP10F' cells for the transformation, but do not add IPTG to the plates. These cells carry the lacIq repressor that represses expression from the lac promoter and so allows cloning of toxic genes. Keep in mind that in the absence of IPTG, blue-white screening cannot be performed.
- Try using Stbl2 cells for the transformation.

What advantages do your Stbl2 cells offer over other cloning strains?

There are other strains available that may function similarly to Stbl2 cells in stabilizing inserts or vectors with repeated DNA sequences. However, one advantage of Stbl2 cells over many similar strains is that they are sensitive to Kanamycin, so you can use Stbl2 to propagate plasmids containing a Kanamycin resistance marker. 

How do you recommend that I prepare my DNA for successful electroporation of E. coli?

For best results, DNA used in electroporation must have a very low ionic strength and a high resistance. A high-salt DNA sample may be purified by either ethanol precipitation or dialysis.

The following suggested protocols are for ligation reactions of 20ul. The volumes may be adjusted to suit the amount being prepared.

Purifying DNA by Precipitation: Add 5 to 10 ug of tRNA to a 20ul ligation reaction. Adjust the solution to 2.5 M in ammonium acetate using a 7.5 M ammonium acetate stock solution. Mix well. Add two volumes of 100 % ethanol. Centrifuge at 12,000 x g for 15 min at 4C. Remove the supernatant with a micropipet. Wash the pellet with 60ul of 70% ethanol. Centrifuge at 12,000 x g for 15 min at room temperature. Remove the supernatant with a micropipet. Air dry the pellet. Resuspend the DNA in 0.5X TE buffer [5 mM Tris-HCl, 0.5 mM EDTA (pH 7.5)] to a concentration of 10 ng/ul of DNA. Use 1 ul per transformation of 20 ul of cell suspension.

Purifying DNA by Microdialysis: Float a Millipore filter, type VS 0.025 um, on a pool of 0.5X TE buffer (or 10% glycerol) in a small plastic container. Place 20ul of the DNA solution as a drop on top of the filter. Incubate at room temperature for several hours. Withdraw the DNA drop from the filter and place it in a polypropylene microcentrifuge tube. Use 1ul of this DNA for each electrotransformation reaction.

Should I increase the heat shock time for my chemically competent cells during the transformation of a larger volume?

The recommended heat shock time does increase slightly with increasing volume of competent cells. For a 50 µl reaction volume, you should heat shock at 42°C for 30 seconds. For 100 µl, 45 seconds is recommended and for 250 µl, 60 seconds. It is important to do a positive control transformation of pUC19 along with transformation of your ligation product to accurately determine your relative efficiency of transformation.

Could the efficiency of my transformation in E. coli improve if I use more than the recommended amount of DNA?

It may be surprising, but in most cases transformation efficiency per µg of DNA will actually decrease when higher amounts of plasmid are transformed in one reaction. While you may see more colonies on your plates, much of the extra plasmid DNA you added will actually be wasted. Competent cells eventually become oversaturated with DNA, and adding more plasmid beyond that level will not result in any additional colonies. For example, when transforming 10 pg plasmid DNA, the efficiency of TOP10 cells is 1.0x10E9 colonies per µg of DNA that you added. If you transform 1 ng all at once, the overall efficiency is likely to decrease to ~1.0x 10E8 colonies per µg, and transforming 1 µg in a single reaction will likely result in efficiency less than 1.0 x 10E6 colonies per µg.

To maximize colony yield, it is better to transform smaller amounts of DNA in multiple reactions rather than adding all of the DNA to one reaction. This is most important when transforming a library, where you ideally want each plasmid to be represented by a colony after transformation.

Can I use E. coli containing an F' plasmid, such as Top 10F', to propagate a vector with the ccdB gene?

While the F' plasmid does contain the ccdA gene that can inhibit or reduce the toxicity of the ccdB gene product, the ccdA expression level is likely to be too low, or inhibition may not be complete, and the bacteria would still be exposed to the ccdB gene product and thus not grow. Therefore, bacterial strains containing the F' plasmid are not recommended as hosts for propagation of ccdB containing vectors.

For propagation of Gateway vectors containing ccdB, we recommend the One Shot ccdB Survival 2 T1R Competent Cells (A10460), which were specifically designed for that purpose. However, please note that these cells are not validated for propagation of other ccdB-containing vectors like the older pZErO plasmids, and in most cases they are not expected to work due to very high levels of ccdB protein expressed in those vectors.

What is the reason for the nupG mutation in the genotypes for TOP10- and DH10B-related E. coli strains?

nupG is a mutation for the transport of nucleosides. The nupG site is next to endA on the chromosome, and when endA was mutated by transposon insertion, the nupG site was unintentionally mutated as well. There are no apparent effects of this mutation on cell function or growth.

References: 1) Nghiem, Y. et al. PNAS 85: 2709-2713. 2) Westh Hansen, S.V. et al. Eur. J. Biochem. 168: 385-391.

Can encapsulated phagemid DNA or M13 phage be used to infect bacteria?

Single-stranded DNA viral particles like M13 require the presence of an F pilus in order to infect E. coli. This criterion is met by TOP10F', DH5? F'IQ, INV?F', Stbl4, OmniMAX2-T1 and DH12S cells. These cells are not traD mutants, which effectively allows the cells to retain the F' episome. Transforming single-stranded DNA can cause a 100- to 1,000-fold reduction in efficiency compared to viral particles.

When should DMSO, formamide, glycerol and other cosolvents be used in PCR?

Cosolvents may be used when there is a failure of amplification, either because the template contains stable hairpin-loops or the region of amplification is GC-rich. Keep in mind that all of these cosolvents have the effect of lowering enzyme activity, which will decrease amplification yield. For more information see P Landre et al (1995). The use of co-solvents to enhance amplification by the polymerase chain reaction. In: PCR Strategies, edited by MA Innis, DH Gelfand, JJ Sninsky. Academic Press, San Diego, CA, pp. 3-16.

Additionally, when amplifying very long PCR fragments (greater than 5 kb) the use of cosolvents is often recommended to help compensate for the increased melting temperature of these fragments.


For Research Use Only. Not for use in diagnostic procedures.