missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin-14-CTP

Catalog No. 19519016
Encompass
Change view
Click to view available options
Quantity:
500 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19519016 500 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19519016 Supplier Invitrogen™ Supplier No. 19519016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Biotin-14-CTP is supplied at 10mM in 10mM Tris-HCl (pH 7.5), 0.1mM EDTA.

A CTP analog with biotin attached at the N 4 -position of the pyrimidine base by a 14-atom linker

Biotin-14-CTP can be efficiently incorporated into RNA transcripts using T7, SP6, or T3 RNA polymerase and the appropriate template in the presence of ATP, GTP, UTP, and Biotin-14-CTP. The biotinylated RNA can be used as a probe in membrane-based or in situhybridizations. The amount of material provided is sufficient for up to 20 transcription reactions (1μg template).

Performance and Quality Testing: Purity is evaluated by reverse-phase HPLC.

DNA and RNA Purification and Analysis, Nucleic Acid Labeling and Oligo Synthesis, RNA Labeling

Order Info

Shipping Condition: Dry Ice

Specifications

Product Type Labeled Nucleotide
Concentration 10 mM
Content And Storage Biotin-14-CTP is supplied at 10 mM in 10 mM Tris-HCl (pH 7.5), 0.1 mM EDTA. Store at -20°C.
Label or Dye Biotin & Other Hapten
Shipping Condition Dry Ice
Quantity 500 nmol
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.