missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 293A Cell Line

Catalog No. R70507
Encompass
Change view
Click to view available options
Quantity:
1 x 107 cells/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R70507 1 x 107 cells/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R70507 Supplier Invitrogen™ Supplier No. R70507

Includes

293A Cell Line

Contains a stably integrated copy of the E1 gene that supplies the E1 proteins (E1a and E1b) required to generate recombinant adenovirus

A subclone of the 293 cell line and has a relatively flat morphology, it facilitates the initial production, amplification and titering of replication-incompetent adenovirus.

  • Flat morphology of cells makes titering procedure easier and suitable for screening adenoviral constructs

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Constitutive Expression, Mammalian Cell Culture, Mammalian Expression, Mammalian Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery, Viral Vaccine Production

Specifications

Content And Storage 1 vial of 1 x 107 cells. Store in liquid nitrogen.
Cell Line 293A
Species Human
Quantity 1 x 107 cells/mL
Product Line ViraPower
At what point can I stop changing the medium on the plates producing adenovirus? It seems that I will be removing adenovirus when replacing the medium.

Most of the adenovirus is contained within the floating cells and is not released into the medium until those cells burst. We recommend changing the medium every 3 days or so until it is obvious that a lot of cells become big and rounded and are detaching from the plastic. Once a cell bursts, the free viruses rapidly infect the neighboring cells. If you're ever worried that you're losing infected cells (and therefore potential virus) in your medium changes, you can always save the medium with the floating cells, freeze/thaw it 3 times and then use a little (maybe 1/10th) and add it back to your culture with fresh media. Or, replace only half of the medium with fresh medium and do this more often than every three days.

Can I use any 293 cells for adenovirus production?

Any 293-derived cell line or other cell line that expresses the E1 proteins may be used to produce adenovirus. In 293A cells (recommended for adenovirus production), "A" stands for "adherent" because the 293A cells (which are just a single-cell clone of regular 293) tend to adhere and form nice flat monolayers in tissue culture dishes. This is why they work so well for plaque assays. Regular 293 cells will not form the same type of monolayers; they exhibit holes and gaps during growth.

Do you recommend a specific FBS for culturing 293A cells? Which plastic plates do you recommend?

We use mycoplasma-tested Gibco FBS (Cat. No. 16000-044) and use the following plasticware for 293A cells:

T175: Fisher Cat. No. 10-126-13; this is a Falcon flask with a 0.2 µm vented plug seal cap.

T75: Fisher Cat. No. 07-200-68; this is a Costar flask with a 0.2 µm vented seal cap.

100 mm plate: Fisher Cat. No. 08-772E; this is a Falcon tissue culture-treated polystyrene plate.

We get excellent adherence on these plates under routine cell culture/maintenance conditions (expect cell lysis in 293A cells when making adenovirus).

Why is it necessary to digest the adenoviral expression construct with Pac I, before transfection into 293A cells?

Before you can transfect your expression clone into 293A cells, you must expose the left and right viral inverted terminal repeats (ITRs) on the vector to allow proper viral replication and packaging. This also removes bacterial sequences (i.e., pUC origin and ampicillin resistance gene). Both pAd/CMV/V5-DEST and pAd/PL-DEST ;vectors contain Pac I restriction sites (see maps on pages 20 and 22 of the manual (http://tools.thermofisher.com/content/sfs/manuals/pad_dest_man.pdf), respectively, for the location of the Pac I sites).

Note: Make sure that your DNA sequence of interest does not contain any Pac I restriction sites. If you are unable to use the Pac I site, you can use the Swa I site.

Does the ViraPower Adenoviral Expression System use an adeno-associated virus?

No. The ViraPower system uses adenovirus type 5. Adenoviruses (Adenoviridae) and adeno-associated viruses (Parvoviridae) are completely different. Adeno-associated viruses are often associated with adenovirus infections, hence the name. Since they are thought to be virtually non-pathogenic, they are attractive vectors for gene therapy. The disadvantage is that they can package only about half the foreign DNA that adenoviruses can.

How does the adenoviral system work? How do I make an adenovirus expressing my gene of interest?

Clone your gene of interest into the pAd/CMV/V5-DEST (or pAd-PL-DEST if you want to use your own promoter). Prior to cloning, if desired, propagate this vector in One Shot ccdB Survival 2 T1R Competent Cells (Cat. No. A10460) as described below. After cloning your gene of interest, propagate in E. coli strain TOP10. pAd/CMV/V5-GW/lacZ is provided as a positive control vector for expression.

Digest recombinant plasmid with Pac I to expose the ITRs (inverted terminal repeats).

Transfect (we recommend Lipofectamine 2000 reagent) E1-containing cells (293A cells) with linear DNA (only 10% of transfected cells will make virus).

Infected cells will ball up, and release virus to surrounding cells, which in turn will be killed and ball up. Look for plaques in the monolayer created by areas cleared by detaching, balled up cells (it takes 8-10 days to see visible plaques from this initial transfection).

Collect a crude viral lysate.

Amplify the adenovirus by infecting 293A producer cells with the crude viral lysate. Harvest virus after 2-3 days when cells ball up. Determine the titer of the adenoviral stock by performing a plaque assay. The virus generated is adenovirus type 5 (subclass C).

Add the viral supernatant to your mammalian cell line of interest to transduce cells.

Assay for recombinant protein of interest.

Once you have your gene of interest in the adenoviral vector, you can simply re-amplify when you need more of the virus. You do not need to repeat cloning steps and transfections each time.

When cloning or propagating DNA with unstable inserts (such as lentiviral DNA containing direct repeats), we recommend using the following modifications to reduce the chance of recombination between direct repeats:
- Select and culture transformants at 25-30 degrees C.
- Do not use "rich" bacterial media as they tend to give rise to a greater number of unwanted recombinants.
-If your plasmid confers chloramphenicol resistance, select and culture transformants using LB medium containing 15-30 µg/mL chloramphenicol in addition to the antibiotic appropriate for selection of your plasmid.

How do I concentrate the lentiviral stock?

Ultracentrifugation is the most commonly used approach and is typically very successful (see Burns et al. (1993) Proc Natl Acad Sci USA 90:8033-8037; Reiser (2000) Gene Ther 7:910-913). Others have used PEG precipitation. Some purification methods are covered by patents issued to the University of California and Chiron.

Adenovirus is concentrated using CsCl density gradient centrifugation (there is a reference for this procedure in our adenovirus manual) or commercially available columns.

Will I get the same transduction efficiency with both lentivirus and adenovirus in the same cell line?

This depends entirely on the target cell. Adenovirus requires the coxsackie-adenovirus receptor (CAR) and an integrin for efficient transduction. Lentivirus (with VSV-G) binds to a lipid in the plasma membrane (present on all cell types). With two totally different mechanisms of entry into the cell, there will always be differences in transduction efficiencies. However, the efficiency of transduction for both viral systems is easily modulated by the multiplicity of infection (MOI) used.

Do you recommend a specific FBS for culture of the 293FT or 293A cells used in the ViraPower kits? What plastic plates do you recommend?

We use mycoplasma-tested Gibco FBS (Cat. No. 16000-044) without any modifications. We have observed that when 293FT cells are cultured in the presence of this FBS following the instructions in the manual, virus production is better than that obtained with many other serum sources.

We use the following plasticware for 293A and 293FT cells:

T175--Fisher Cat. No. 10-126-13; this is a Falcon flask with 0.2 µm vented plug seal cap.

T75--Fisher Cat. No. 07-200-68; this is a Costar flask with 0.2 µm vented seal cap.

100 mm plate--Fisher Cat. No. 08-772E; this is a Falcon tissue culture-treated polystyrene plate

We get excellent adherence on these plates under routine cell culture/maintenance conditions (expect cell lysis in 293A cells when making adenovirus).

How should I store lentivirus, adenovirus and viral vectors?

Viral vectors:
Store lentiviral and adenoviral expression vectors (plasmid DNA) at -20 degrees C. Due to their relatively large sizes, we do not recommend storing these vectors at -80 degrees C, as the vector solutions will completely freeze and too many freeze thaws from -80 degrees C will affect the cloning efficiency. At -20 degrees C, the vectors will be stable but will not freeze completely. Glycerol stocks of vectors transformed into bacteria should always be stored at -80 degrees C.

Virus:
Both adenovirus and lentivirus particles should be aliquoted immediately after production and stored at -80 degrees C.

Lentivirus is more sensitive to storage temperature and to freeze/thaw than adenovirus and should be handled with care. Adenovirus can typically be frozen/thawed up to 3 times without loss of titer, while lentivirus can lose up to 5% or more activity with each freeze/thaw. It is recommended to aliquot your virus into small working volumes immediately after production, freeze at -80 degrees C, and then thaw just one aliquot for titering. This way, every time you thaw a new aliquot it should be the same titer as your first tube.

Adenovirus particles can be kept overnight at 4 degrees C if necessary, but it is best to avoid this. Viruses will be most stable at -80 degrees C.

When stored properly, viral stocks should maintain consistent titer and be suitable for use for up to one year. After long-term storage, we recommend re-titering your viral stocks before use.

What are the safety issues associated with the use of your viral systems?

Both the lentiviral and adenoviral systems should be used following Biosafety Level 2 (BSL-2). We recommend strict adherence to all CDC guidelines for BSL-2 (as well as institutional guidelines). Thermo Fisher Scientific has also engineered specific safety features into the lentiviral system.

Consult the "Biosafety in Microbiological and Biomedical Laboratories" publication (www.cdc.gov, published by the CDC in the USA, describes BSL-2 handling) and the "Laboratory Biosafety Guidelines" publication (www.phac-aspc.gc.ca, published by the Centre for Emergency Preparedness and Response in Canada) for more information on safe handling of various organisms and the physical requirements for facilities that work with them.

How do I know whether to choose lentivirus or adenovirus for viral expression?

If you're interested in stable integration and selection, choose the lentiviral system. We offer both a Directional TOPO (D-TOPO) and Gateway version of the kit to provide flexibility in the cloning of the gene of interest.

If you're looking for transient gene expression, choose the adenoviral system. We offer the Gateway cloning method for this product. It should be noted, however, that gene expression from both systems is typically detected within 24-48 hours of transduction, so both systems can be used for experiments of a transient nature. The main difference is that lentivirus integrates into the host genome and adenovirus does not. Higher viral titers are achieved with the adenovirus.

What are the packaging limits for lentivirus and adenovirus? Can a 9 kb fragment be packaged into either?

No, neither lentivirus nor adenovirus can take an insert as large as 9 Kb. Lentiviral packaging limits are around 6 kb and adenoviral packaging limits are around 7-7.5 kb. Above that, no virus is made.

For lentivirus, titers will generally decrease as the size of the insert increases. We have effectively packaged inserts of 5.2 kb with good titer (approx. 0.5 x 10^5 cfu/mL). The size of the wild-type HIV-1 genome is approximately 10 kb. Since the size of the elements required for expression from pLenti vectors add up to approximately 4-4.4 kb, the size of your gene of interest should theoretically not exceed 5.6-6 kb for efficient packaging (see below for packaging limits for individual vectors).
pLenti4/V5-DEST vector: 6 kb
pLenti6/V5-DEST vector: 6 kb
pLenti6/V5/D-TOPO vector: 6 kb
pLenti6/UbC/V5-DEST vector: 5.6 kb

For adenovirus, the maximum packagable size is approximately 7-7.5 Kb (see below for packaging limits for individual vectors).
pAd/CMV/V5-DEST vector: 6 kb
pAd/PL-DEST vector: 7.5 kb


For Research Use Only. Not for use in diagnostic procedures.