missing translation for 'onlineSavingsMsg'
Learn More

Promega Intracellular TE Nano-Glo Substrate/Inhibitor, 1,000 assays

Catalog No. PRN2160
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN2160 1000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRN2160 Supplier Promega Supplier No. N2160*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Intracellular TE Nano-Glo Substrate/Inhibitor is a supplement for NanoBRET™ Target Engagement Assays.

  • Dedicated Reagents for Intracellular NanoBRET™ TE Assays
  • Combination of NanoLuc(R) Substrate and Extracellular NanoLuc(R) Inhibitor
  • Ensures measurements originate from inside intact cells
  • For use with NanoBRET™ Intracellular Target Engagment Tracers

Specifications

Content And Storage -30°C to -10°C
Quantity 1000 Assays