missing translation for 'onlineSavingsMsg'
Learn More

Internal Standard, SPEX CertiPrep™

Numéro de catalogue. CLPSI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
CLPSI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CLPSI Fournisseur SPEX CertiPrep Group LLC Code fournisseur CLPSI
Il en reste null

Comprend

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,4-Dichlorobenzene-d43855-82-14000μg/mL
Acenaphthene-d1015067-26-24000μg/mL
Chrysene-d121719-03-54000μg/mL
Naphthalene-d81146-65-24000μg/mL
Perylene-d121520-96-34000μg/mL
Phenanthrene-d101517-22-24000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Spécifications

Chemical Name or Material Internal Standard
Packaging 2mL amber prescored glass ampule
Name Note EPA Method 625, SW-846 Method 8270C, CLP Methods
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Methods For US EPA Methods 625, 8270C, CLP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix Methylene Chloride
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 4000μg/mL

Product intended for laboratory use only