missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interference Check Standard 2 For ICP/MS, SPEX CertiPrep™
Description
Comprend
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ag | 10μg/mL |
| As | 10μg/mL |
| Cd | 10μg/mL |
| Co | 10μg/mL |
| Cr | 10μg/mL |
| Cu | 10μg/mL |
| Mn | 10μg/mL |
| Ni | 10μg/mL |
| Zn | 10μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Spécifications
Spécifications
| Chemical Name or Material | Interferents Check Solution B2 |
| Packaging | Triple-leached LDPE bottle |
| Name Note | For SW-846 |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | AA, ICP |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | 2% HNO3 |
| Quantity | 125 mL |
| Grade | ICP/ICP-MS |
| Afficher plus de résultats |
Product intended for laboratory use only