missing translation for 'onlineSavingsMsg'
Learn More

Instrument Calibration Standard 2, Ricca Chemical

Catalog No. INSTCL2100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
INSTCL2100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. INSTCL2100 Supplier Ricca Chemical Company Supplier No. INSTCL2100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1310-73-2 , 7439-96-5 , 7440-02-0 , 7697-37-2 , 7440-22-4 , 7732-18-5; 7440-66-6
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Packaging Amber Poly Bottle
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Multi-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Instrument Calibration Standard #2
ShelfLife 12 months
Recommended Storage 15°C - 30°C