missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Expert pH Electrodes

A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.

$793.52 - $1,344.79

Specifications

Material PEEK
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Filling Solution Type Xerolyt Polymer
Internal Reference ARGENTHAL
View More Specs

Products 10
Catalog Number Mfr. No. Connector Types Includes Length (Metric) Cable Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types Includes Length (Metric) Cable Price Quantity & Availability  
01911463
View Documents Promotion Details
METTLER TOLEDO™
51343104
Multi-Pin Temperature Probe (NTC30) No Cable
Each for $942.77
Only null left
 
01911460
View Documents Promotion Details
METTLER TOLEDO™
51343100
S7 Electrode only No Cable
Each for $793.52
Only null left
 
01911462
View Documents Promotion Details
METTLER TOLEDO™
51343103
DIN 19262, 4 mm Banana Plug Pt1000 Temperature Probe 1.2 m
Each for $1,057.58
Only null left
 
01911465
View Documents Promotion Details
METTLER TOLEDO™
51343102
BNC, RCA ISM Temperature Probe 2 m
Each for $1,051.53
Only null left
 
01911464
View Documents Promotion Details
METTLER TOLEDO™
51343105
Multi-Pin Pt1000 Temperature Probe No Cable
Each for $942.77
Only null left
 
01911461
View Documents Promotion Details
METTLER TOLEDO™
51343101
BNC, RCA Temperature Probe 1.2 m
Each for $1,011.27
Only null left
 
01910552
View Documents Promotion Details
METTLER TOLEDO™
51340288
BNC, RCA IP67 Temperature Probe 2 m
Each for $1,344.79
Only null left
 
01917140
View Documents Promotion Details
METTLER TOLEDO™
51344102
BNC, RCA IP67 ISM Temperature Probe 1.8 m
Each for $1,008.74
Only null left
 
01917141
View Documents Promotion Details
METTLER TOLEDO™
51344104
BNC, RCA IP67 ISM Temperature Probe 10 m
Each for $1,112.33
Only null left
 
01915108
View Documents Promotion Details
METTLER TOLEDO™
30014096
BNC, RCA ISM Temperature Probe 1.2 m
Each for $819.53
Only null left
 
Description

Description

Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Features a PEEK plastic shaft for harsh environments and requires no maintenance
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • General Lab
  • Beverages
  • Waste water
  • Suspended solids
  • Environmental
  • TRIS compatible

Accessories & Care

  • Open Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications

Specifications

PEEK
0°C to 100°C
ARGENTHAL
120 mm
12 mm
0 to 14
Xerolyt Polymer
U Glass
0.47 in.
Videos
SDS
Documents

Documents