Learn More
METTLER TOLEDO™ InLab™ Expert pH Electrodes
A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.
Supplier: METTLER TOLEDO™ 51343103
Description
Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.
- For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
- Features a PEEK plastic shaft for harsh environments and requires no maintenance
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- General Lab
- Beverages
- Waste water
- Suspended solids
- Environmental
- TRIS compatible
Accessories & Care
- Open Junction and non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Chemical Identifiers
| PEEK | |
| 0°C to 100°C | |
| ARGENTHAL | |
| 120 mm | |
| 12 mm |
| 0 to 14 | |
| Xerolyt Polymer | |
| U Glass | |
| 0.47 in. |
Specifications
| DIN 19262, 4 mm Banana Plug | |
| PEEK | |
| 0°C to 100°C | |
| Xerolyt Polymer | |
| pH Meter | |
| ARGENTHAL | |
| InLab Expert DIN | |
| <20 sec. | |
| 0°F to 212°F | |
| 4.7 in. | |
| 120 mm | |
| 0.47 in. |
| Dual | |
| 0 to 14 | |
| Combination pH Electrode, Integrated Pt1000 Temperature Probe | |
| Emulsions, suspensions, effluent water and solutions containing Proteins and Sulphides | |
| Pt1000 Temperature Probe | |
| U Glass | |
| PT1000 | |
| FRISCOLYT-B | |
| 0°C to 100°C | |
| 47 in. | |
| 1.2 m | |
| 12 mm |