missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Expert pH Electrodes

A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.

Supplier:  METTLER TOLEDO™ 51343100

 View more versions of this product

Catalog No. 01911460


Only null left

Description

Description

Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Features a PEEK plastic shaft for harsh environments and requires no maintenance
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • General Lab
  • Beverages
  • Waste water
  • Suspended solids
  • Environmental
  • TRIS compatible

Accessories & Care

  • Open Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications

Chemical Identifiers

PEEK
0°C to 100°C
ARGENTHAL
120 mm
12 mm
0 to 14
Xerolyt Polymer
U Glass
0.47 in.

Specifications

S7
PEEK
0°C to 100°C
Xerolyt Polymer
S7 to BNC (30281915)
ARGENTHAL
InLab Expert
FRISCOLYT-B
0°C to 100°C
120 mm
0.47 in.
Dual
0 to 14
Combination pH Electrode
Emulsions, suspensions, effluent water and solutions containing Proteins and Sulphides
Electrode only
U Glass
<20 sec.
0°F to 212°F
4.7 in.
No Cable
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions