missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ Ingenio™ Electroporation Solution
Proprietary Ingenio™ Electroporation Solution facilitates efficient and reliable delivery of nucleic acids to eukaryotic cells resistant to chemical transfection.
Supplier: Mirus Bio™ MIR 50117
Description
- Broad spectrum solution supports high efficiency electroporation of hard to transfect cells with minimal toxicity
- Compatible with all electroporation devices: Use with your existing system; no need to purchase additional specialized equipment
- Replaces standard electroporation solutions including phosphate buffered saline and serum-free media
- High cell viability: Minimize the risk of introducing experimental bias due to toxicity-induced cellular changes
Identifiants chimiques
| Electroporation | |
| Electroporation Solution | |
| 4°C |
| All electroporation device | |
| Sterile | |
| Exponential Decay Wave |
Spécifications
| Electroporation | |
| Electroporation Solution | |
| 12.5 mL | |
| 4°C |
| All electroporation device | |
| 100 Reactions | |
| Sterile | |
| Exponential Decay Wave |