missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ Ingenio™ Electroporation Solution
Proprietary Ingenio™ Electroporation Solution facilitates efficient and reliable delivery of nucleic acids to eukaryotic cells resistant to chemical transfection.
$663.92 - $1,843.39
Specifications
| For Use With (Application) | Electroporation |
|---|---|
| For Use With (Equipment) | All electroporation device |
| Product Type | Electroporation Solution |
| Sterility | Sterile |
| Content And Storage | 4°C |
| Catalog Number | Mfr. No. | Quantity | Volume (Metric) | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Volume (Metric) | Price | Quantity & Availability | ||||
|
MIR50114
|
Mirus Bio™
MIR 50114 |
50 Reactions | 12.5 mL |
Each for $1,054.46
|
|
||||
|
MIR50111
|
Mirus Bio™
MIR 50111 |
25 Reactions | 6.25 mL |
Each for $663.92
|
|
||||
|
MIR50117
|
Mirus Bio™
MIR 50117 |
100 Reactions | 12.5 mL |
Each for $1,843.39
|
|
||||
Description
- Broad spectrum solution supports high efficiency electroporation of hard to transfect cells with minimal toxicity
- Compatible with all electroporation devices: Use with your existing system; no need to purchase additional specialized equipment
- Replaces standard electroporation solutions including phosphate buffered saline and serum-free media
- High cell viability: Minimize the risk of introducing experimental bias due to toxicity-induced cellular changes
Specifications
| Electroporation | |
| Electroporation Solution | |
| 4°C |
| All electroporation device | |
| Sterile | |
| Exponential Decay Wave |