missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Industrial Grade Low-Lint Nylon Inspection Gloves

Numéro de catalogue. p-7230587
Change view
Click to view available options
Gender:
Men's
Women's
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Gender
19075288 Men's
19075289 Women's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 19075288 Fournisseur Fisherbrand™ Code fournisseur 98700
Il en reste null

Heavy-weight gloves are constructed of 40-denier nylon tricot fabric and can be worn alone, or used as a glove liner.

  • Heavyweight fabric constructed in a full-fashion pattern
  • Easy size identification by color-coded hem on zig-zag stitched cuff
  • Provides limited heat protection
  • Moisture absorbent
  • Ambidextrous

Spécifications

Coating Material Uncoated
Color White
Disposable Limited-use
Grade Industrial
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size Large
Sterility Non-sterile
For Use With (Application) Inspection, Electronics Assembly
Gender Men's
Glove Cuff Style Beaded
Hem Color Blue
Length (English) 9.3 in.
Length (Metric) 23.7 cm
Afficher plus de résultats Afficher moins de résultats