missing translation for 'onlineSavingsMsg'
Learn More

Indium ICP-MS Standard, 1000 ppm In in 3% HNO3, Ricca Chemical

Catalog No. MSIN1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSIN1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSIN1KN100 Supplier Ricca Chemical Company Supplier No. MSIN1KN100

Chemical Identifiers

CAS 7440-74-6 , 7697-37-2 , 7732-18-5
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
IUPAC Name indium
SMILES [In]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm In
CAS Min % 90.0, 8.1, 0.09
CAS Max % 93.0, 8.43, 0.09
Equivalence 1 mL = 1 mg In
Physical Form Liquid
Color Colorless
Quantity 100 mL
Chemical Name or Material Indium ICP-MS Standard