missing translation for 'onlineSavingsMsg'
Learn More

ACCU-SCOPE™ Immersion Oil

Catalog No. S00158
Change view
Click to view available options
Viscosity:
150 cSt
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Viscosity
S00158 150 cSt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00158 Supplier ACCU-SCOPE™ Supplier No. 3205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Immersion Oil - Type A, 1/4 oz. Cargille

Cargille, Type A immersion oil

General purpose use

Nearly fluorescence free

Refractive Index @ 23°C: e Line 9546.1 nm) - 1.5180

Viscosity: 150 cSt ± 10% @ 23°C (Low)

Specifications

Type Immersion Oil Type A
Viscosity 150 cSt