missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Imidazole, 0.5M buffer soln., pH 7.5

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

$188.89 - $294.81

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAJ63496AE
View Documents
Thermo Scientific Chemicals
J63496AE
100 mL
Each for $188.89
Only null left
 
AAJ63496AK
View Documents
Thermo Scientific Chemicals
J63496AK
250 mL
Each for $294.81
Only null left
 
Description

Description

Imidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

288-32-4
68.08
RAXXELZNTBOGNW-UHFFFAOYSA-N
N1C=CN=C1
C3H4N2
MFCD00005183
1H-imidazole

Specifications

288-32-4
7.5
100 mL
MFCD00005183
N1C=CN=C1
68.08
0.5 M
Liquid
C3H4N2
RAXXELZNTBOGNW-UHFFFAOYSA-N
1H-imidazole
Imidazole
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only