Learn More
Thermo Scientific Chemicals Imidazole, 0.5M buffer soln., pH 7.5
CAS: 288-32-4 | C3H4N2 | 68.08 g/mol
Supplier: Thermo Scientific Chemicals J63496AE
Description
Imidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsImidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.
Solubility
Fully miscible.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Safety and Handling
Recommended Storage : Keep cold
RUO – Research Use Only