accessibility menu, dialog, popup

UserName

HNN

CAS RN 288-32-4

IUPAC Name: 1H-imidazole
Synonyms: imidazole|glyoxaline|iminazole|glyoxalin|imutex|cu-imidazole|glioksal|al-imidazole|pyrro(b)monazole|miazole
Molecular Weight (g/mol): 68.08
Molecular Formula: C3H4N2
InChi Key: RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES: N1C=CN=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)