Learn More
Invitrogen™ Mouse IL-28 ELISA Kit
Description
Comprend
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-28
Biotin-Conjugate anti-mouse IL-28 monoclonal antibody
Streptavidin-HRP
Mouse IL-28 Standard lyophilized, 4000pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x
The Mouse Interleukin-28A (Ms IL-28A) ELISA quantitates Ms IL-28A in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-28A. Principle of the method The Mouse IL-28A solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Spécifications
Spécifications
| Accession Number | Q4VK74 |
| Assay Range | 31.3 to 2,000 pg/mL |
| Assay Sensitivity | 12.85 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 330496 |
| Afficher plus de résultats |