missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat IL-12 p40 ELISA Kit

ELISA

$916.00 - $1,406.00

Spécifications

Accession Number Q9R103
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity <3 pg/mL
Conjugate HRP
Product Type ELISA
voir les spécifications

Comprend: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
KRC0121
Encompass
Afficher les documents Détails de la promotion
Invitrogen™
KRC0121
96 Tests
chaque for $916.00
Il en reste null
 
KRC0122
Encompass
Afficher les documents Détails de la promotion
Invitrogen™
KRC0122
2 x 96 Tests
chaque for $1,406.00
Il en reste null
 
Description

Description

The Rat Interleukin 12+p40 (Rt IL-12+p40) ELISA quantitates Rt IL-12+p40 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt IL-12+p40. Principle of the method The Rat IL-12+p40 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IL12B (IL-12 p40) encodes a subunit of interleukin 12, a cytokine that acts on T and natural killer cells, and has a broad array of biological activities. Interleukin 12 is a disulfide-linked heterodimer composed of the 40 kDa cytokine receptor like subunit encoded by this gene, and a 35 kDa subunit encoded by IL12A. This cytokine is expressed by activated macrophages that serve as an essential inducer of Th1 cells development. This cytokine has been found to be important for sustaining a sufficient number of memory/effector Th1 cells to mediate long-term protection to an intracellular pathogen. Overexpression of this gene was observed in the central nervous system of patients with multiple sclerosis (MS), suggesting a role of this cytokine in the pathogenesis of the disease. The promoter polymorphism of this gene has been reported to be associated with the severity of atopic and non-atopic asthma in children.

Order Info

Shipping Condition: Wet ice

Spécifications

Spécifications

Q9R103
<3 pg/mL
ELISA
Rat
Colorimetric Microplate Reader
Il12a, Il12b
4.2
HRP
RUO
2°C to 8°C
1 hrs 20 mins
15.6 to 1000 pg/mL
HRP
Plasma, Serum, Supernatant
Colorimetric
64546
5.1%
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
<3 pg/mL
Plasma 10 μL, Serum 10 μL, Supernatant 100 μL
IL-12 p40
4 hrs
Videos
FDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.