Learn More
QuidelOrtho ID-Micro Typing System™ Gel Test™ Cards
Delivered with gel-filled microtubes infused with pre-measured reagents
Spécifications
| Certifications/Compliance | cGMP |
|---|---|
| For Use With (Application) | For broad range of Blood bank applications, Antibody screening and identification, ABO Blood grouping and Rh phenotyping, compatibility testing and reverse Serum grouping and Antigen typing |
| Includes | Gel-filled microtubes |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Type | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Type | Prix | Quantity & Availability | ||||
OKMTS081115
|
QuidelOrtho
MTS081115 |
100 Cards | ID-Micro Typing System™ A/B/D Monoclonal Grouping Card |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
OKMTS081001
|
QuidelOrtho
MTS081001 |
20 Cards | ID-Micro Typing System™ MTS Monoclonal Control Card |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
OKMTS085014
|
QuidelOrtho
MTS085014 |
100 Cards | ID-Micro Typing System™ Buffered Gel Card |
N/A
|
N/A | ||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
OKMTS084024
|
QuidelOrtho
MTS084024 |
100 Cards | ID-Micro Typing System™ MTS Anti-IgG Card |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
OKMTS084014
|
QuidelOrtho
MTS084014 |
20 Cards | ID-Micro Typing System™ Anti-IgG,-C3d Card |
N/A
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
QuidelOrtho ID-Micro Typing System is suitable for a broad range of blood bank applications, including antibody screening and identification, ABO blood grouping and Rh phenotyping, compatibility testing, and reverse serum grouping and antigen typing, all using proven serologic methods. Gel Test cards are prefilled cards containing gel mixed with the appropriate reagent. Simply pipet, incubate, spin and read: results are ready to report, without the need for cell washing, red cell resuspension or tube shaking.
Standardization
- Standardized methods ensure uniform results from tech-to-tech and laboratory-to-laboratory
- Enhanced cGMP compliance is attained through standardized procedures and simplified training
Safety
- Opportunities for error are reduced by stable and clear endpoints that provide objective, consistent results
- Ease of Use
- Gel testing is easy to learn, use, and teach: the ideal technology for all shifts and all levels of blood banking experience
- Eliminates washing, shaking, and the need to add control cells
Cost-effectiveness
- Gel testing requires fewer steps than tube testing, dramatically reducing hands-on time, with less waste and costly re-work
- Well-balanced sensitivity and specificity means less time and fewer resources spent on clinically unimportant antibodies
Spécifications
| cGMP | |
| For broad range of Blood bank applications, Antibody screening and identification, ABO Blood grouping and Rh phenotyping, compatibility testing and reverse Serum grouping and Antigen typing | |
| Gel-filled microtubes |