Learn More
QuidelOrtho ID-Micro Typing System™ Gel Test™ Cards
Catalog No. OKMTS081115
Encompass
Change view
Click to view available options
Quantity:
100 Cards
20 Cards
Type:
ID-Micro Typing System™ A/B/D Monoclonal Grouping Card
ID-Micro Typing System™ Anti-IgG,-C3d Card
ID-Micro Typing System™ Buffered Gel Card
ID-Micro Typing System™ MTS Anti-IgG Card
ID-Micro Typing System™ MTS Monoclonal Control Card
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
OKMTS081115 100 Cards ID-Micro Typing System™ A/B/D Monoclonal Grouping Card
OKMTS081001 20 Cards ID-Micro Typing System™ MTS Monoclonal Control Card
OKMTS085014 100 Cards ID-Micro Typing System™ Buffered Gel Card
OKMTS084024 100 Cards ID-Micro Typing System™ MTS Anti-IgG Card
OKMTS084014 20 Cards ID-Micro Typing System™ Anti-IgG,-C3d Card
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. OKMTS081115 Supplier QuidelOrtho Supplier No. MTS081115
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Delivered with gel-filled microtubes infused with pre-measured reagents
QuidelOrtho ID-Micro Typing System is suitable for a broad range of blood bank applications, including antibody screening and identification, ABO blood grouping and Rh phenotyping, compatibility testing, and reverse serum grouping and antigen typing, all using proven serologic methods. Gel Test cards are prefilled cards containing gel mixed with the appropriate reagent. Simply pipet, incubate, spin and read: results are ready to report, without the need for cell washing, red cell resuspension or tube shaking.
Standardization
  • Standardized methods ensure uniform results from tech-to-tech and laboratory-to-laboratory
  • Enhanced cGMP compliance is attained through standardized procedures and simplified training
Safety
  • Opportunities for error are reduced by stable and clear endpoints that provide objective, consistent results
  • Ease of Use
  • Gel testing is easy to learn, use, and teach: the ideal technology for all shifts and all levels of blood banking experience
  • Eliminates washing, shaking, and the need to add control cells
Cost-effectiveness
  • Gel testing requires fewer steps than tube testing, dramatically reducing hands-on time, with less waste and costly re-work
  • Well-balanced sensitivity and specificity means less time and fewer resources spent on clinically unimportant antibodies
Specifications
Certifications/Compliance cGMP
Type ID-Micro Typing System™ A/B/D Monoclonal Grouping Card
For Use With (Application) For broad range of Blood bank applications, Antibody screening and identification, ABO Blood grouping and Rh phenotyping, compatibility testing and reverse Serum grouping and Antigen typing
Includes Gel-filled microtubes
Quantity 100 Cards