missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind™ Flex Solution Kit

Numéro de catalogue. p-5013595
Encompass
Change view
Click to view available options
Quantity:
1 kit
4 kits
Product Type:
Flex Solution Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Product Type
SLF2020 1 kit Flex Solution Kit
SLF2020X4 4 kits Flex Solution Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. SLF2020 Fournisseur Invitrogen™ Code fournisseur SLF2020
Il en reste null

Detection kit for use with iBind™ Flex Western System

iBind™ Flex Solution is a proprietary formulation compatible with downstream fluorescent detection/chemiluminescent detection. It serves as a combined blocking, washing and antibody diluent solution to process up to one midi blot, two mini blots, or six vertically cut strip blots when used with the iBind™ Flex Western Device and iBind™ Flex Cards.

Each kit contains reagents sufficient for 10 midi blots or 20 mini blots

Fluorescent Detection (FD) Solution (Contents)

  • iBind Flex FD 5X Buffer
  • iBind Flex 100X Additive
  • iBind Flex FD 10% SDS

Chemiluminescent Solution (Contents)

  • iBind Flex 5X Buffer
  • iBind Flex 100X Additive

Store at 2° to 8°C

Spécifications

Product Line iBind
Membrane Compatibility Nitrocellulose, PVDF
Detection Method Colorimetric, Chromogenic, Chemiluminescence
Product Type Flex Solution Kit
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
No. of Reactions 10 midi blots or 20 mini blots
For Use With (Equipment) iBind Flex Western Device
Content And Storage • iBind Flex 5X Buffer
• iBind Flex 100X Additive

Store at 2–8°C.
Kit Contents iBind™ Flex 5X buffer, iBind™ Flex 100X additive
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.