missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyflutamide 98.0+%, TCI America™
$466.02 - $1392.44
Chemical Identifiers
| CAS | 52806-53-8 |
|---|---|
| Molecular Formula | C11H11F3N2O4 |
| Molecular Weight (g/mol) | 292.214 |
| MDL Number | MFCD00563126 |
| InChI Key | YPQLFJODEKMJEF-UHFFFAOYSA-N |
| Synonym | 2-Hydroxy-4′-nitro-3′-(trifluoromethyl)isobutyranilide, 2-Hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide, 2-Hydroxyflutamide |
| PubChem CID | 91649 |
| ChEBI | CHEBI:43064 |
| IUPAC Name | 2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide |
| SMILES | CC(C)(C(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(F)(F)F)O |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
H160025MG
|
TCI America
H160025MG |
25 mg |
Each for $466.02
|
|
|||||
|
H1600100MG
|
TCI America
H1600100MG |
100 mg |
Each for $1,392.44
|
|
|||||
Chemical Identifiers
| 52806-53-8 | |
| 292.214 | |
| YPQLFJODEKMJEF-UHFFFAOYSA-N | |
| 91649 | |
| 2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide |
| C11H11F3N2O4 | |
| MFCD00563126 | |
| 2-Hydroxy-4′-nitro-3′-(trifluoromethyl)isobutyranilide, 2-Hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide, 2-Hydroxyflutamide | |
| CHEBI:43064 | |
| CC(C)(C(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(F)(F)F)O |
Specifications
| 52806-53-8 | |
| White-Yellow | |
| MFCD00563126 | |
| 3077 | |
| YPQLFJODEKMJEF-UHFFFAOYSA-N | |
| 2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide | |
| 91649 | |
| 292.21 | |
| Crystalline Powder |
| 152°C | |
| C11H11F3N2O4 | |
| 25 mg | |
| 2-Hydroxy-4′-nitro-3′-(trifluoromethyl)isobutyranilide, 2-Hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide, 2-Hydroxyflutamide | |
| CC(C)(C(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(F)(F)F)O | |
| 292.214 | |
| CHEBI:43064 | |
| ≥98.0% (HPLC) | |
| Hydroxyflutamide |
Safety and Handling
TSCA : No