accessibility menu, dialog, popup

UserName

CH3H3COHONHFFFNOO

CAS RN 52806-53-8

IUPAC Name: 2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
Synonyms: hydroxyflutamide
Molecular Weight (g/mol): 292.21
Molecular Formula: C11H11F3N2O4
InChi Key: YPQLFJODEKMJEF-UHFFFAOYSA-N
SMILES: CC(C)(O)C(=O)NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results