missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyflutamide 98.0+%, TCI America™
Supplier: TCI America H1600100MG
Specifications
| Hydroxyflutamide | |
| 152°C | |
| C11H11F3N2O4 | |
| 100 mg | |
| 2-Hydroxy-4′-nitro-3′-(trifluoromethyl)isobutyranilide, 2-Hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide, 2-Hydroxyflutamide | |
| CC(C)(C(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(F)(F)F)O | |
| 292.214 | |
| CHEBI:43064 | |
| ≥98.0% (HPLC) |
| 52806-53-8 | |
| White-Yellow | |
| MFCD00563126 | |
| 3077 | |
| YPQLFJODEKMJEF-UHFFFAOYSA-N | |
| 2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide | |
| 91649 | |
| 292.21 | |
| Crystalline Powder |
Chemical Identifiers
| 52806-53-8 | |
| 292.214 | |
| YPQLFJODEKMJEF-UHFFFAOYSA-N | |
| 91649 | |
| 2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide |
| C11H11F3N2O4 | |
| MFCD00563126 | |
| 2-Hydroxy-4′-nitro-3′-(trifluoromethyl)isobutyranilide, 2-Hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide, 2-Hydroxyflutamide | |
| CHEBI:43064 | |
| CC(C)(C(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(F)(F)F)O |
Safety and Handling
TSCA : No