missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyflutamide 98.0+%, TCI America™

Supplier:  TCI America H160025MG

 View more versions of this product

Catalog No. H160025MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hydroxyflutamide
152°C
C11H11F3N2O4
25 mg
2-Hydroxy-4′-nitro-3′-(trifluoromethyl)isobutyranilide, 2-Hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide, 2-Hydroxyflutamide
CC(C)(C(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(F)(F)F)O
292.214
CHEBI:43064
≥98.0% (HPLC)
52806-53-8
White-Yellow
MFCD00563126
3077
YPQLFJODEKMJEF-UHFFFAOYSA-N
2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
91649
292.21
Crystalline Powder

Chemical Identifiers

52806-53-8
292.214
YPQLFJODEKMJEF-UHFFFAOYSA-N
91649
2-hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
C11H11F3N2O4
MFCD00563126
2-Hydroxy-4′-nitro-3′-(trifluoromethyl)isobutyranilide, 2-Hydroxy-2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide, 2-Hydroxyflutamide
CHEBI:43064
CC(C)(C(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(F)(F)F)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions