missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Sulfide Water, Solution, Spectrum™ Chemical

$675.81 - $1,152.41

Specifications

Chemical Name or Material Hydrogen Sulfide Water
CAS 7783-06-4 , 7732-18-5
CAS Max % 0.4,99.6
Grade Chemical Solution
Packaging Amber Glass Bottle
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610965
View Documents
Spectrum Chemical Mfg Cor
H158500ML
500 mL Amber Glass Bottle N/A
Only null left
 
18610964
View Documents
Spectrum Chemical Mfg Cor
H1581LT
1 L Amber Glass Bottle N/A
Only null left
 
Description

Description

Specifications

Specifications

Hydrogen Sulfide Water
0.4,99.6
Amber Glass Bottle
7783-06-4 , 7732-18-5
Chemical Solution
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.