accessibility menu, dialog, popup

UserName

CAS RN 7783-06-4

IUPAC Name: hydrogen sulfide
Synonyms: hydrogen sulfide super six hydrogen sulphide sulfur atom imber sewer gas sour gas greggio hydrogen sulfide (H2S) hydrogen sulfide (h2s)
Molecular Weight (g/mol): 34.08
Molecular Formula: H2S
InChi Key: RWSOTUBLDIXVET-UHFFFAOYSA-N
SMILES: S
Grade 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results