missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Sulfide Water, Solution, Spectrum™ Chemical

Catalog No. 18610965
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610965 500 mL Amber Glass Bottle
18610964 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610965 Supplier Spectrum Chemical Mfg Cor Supplier No. H158500ML
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Hydrogen Sulfide Water
CAS 7783-06-4, 7732-18-5
CAS Max % 0.4, 99.6
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.