missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen PerOxide Solution, 3 wt% in H2O, Honeywell Fluka™

Numéro de catalogue. p-7072071
Change view
Click to view available options
Quantity:
25 mL
500 mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
6000795 25 mL
6000797 500 mL
6000796 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 6000795 Fournisseur Honeywell Chemicals Code fournisseur 32338125ML
Il en reste null

Contains ∼200 ppm acetanilide as stabilizer, 3 wt. % in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Identifiants chimiques

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Spécifications

UN Number NONH for all modes of transport
Beilstein 3587191
Density 1g/mL at 25°C
Formula Weight 34.01g/mol
Vapor Pressure 23.3mmHg (30°C)
Vapor Density 1.1 (vs air)
Quantity 25 mL
Chemical Name or Material Hydrogen Peroxide Solution