missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, Certified, 3.0% (w/w) to 3.5% (w/w), LabChem™

Numéro de catalogue. LC154501
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
LC154501 500 mL Poly Bottle
LC154502 1 L Poly Bottle
LC154504 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. LC154501 Fournisseur LabChem Code fournisseur LC154501
Il en reste null

Identifiants chimiques

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Spécifications

Color Colorless
CAS Max % 97, 3
Physical Form Liquid
Quantity 500 mL
Linear Formula H2O2
Solubility Information Soluble in water
Formula Weight 34.01
Percent Purity 3.0 to 3.5% (w/w)
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Oxygen
Density 1g/mL
Chemical Name or Material Hydrogen Peroxide
Afficher plus de résultats Afficher moins de résultats
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Recommended Storage Room Temperature