missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, 3% (w/w), Stabilized Reagent Grade, Ricca Chemical
$96.81 - $2561.09
Chemical Identifiers
| CAS | 7722-84-1 |
|---|---|
| Molecular Formula | H2O2 |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
| PubChem CID | 784 |
| ChEBI | CHEBI:16240 |
| IUPAC Name | peroxol |
| SMILES | OO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
3819-16
|
Ricca Chemical Company
381916 |
500 mL | Amber Poly Bottle |
Each for $96.81
|
|
||||
|
3819-32
|
Ricca Chemical Company
381932 |
1 L | Amber Poly Bottle |
Each for $175.44
|
|
||||
|
3819-1
|
Ricca Chemical Company
38191 |
4 L | Black Poly Bottle |
Each for $347.38
|
|
||||
|
RIC01001SOL
|
Ricca Chemical Company
38195HP |
20 L | Hard Pack |
Each for $1,961.94
|
|
||||
|
381955
|
Ricca Chemical Company
381955 |
55 gal. | Poly Drum |
Each for $2,561.09
|
|
||||
Chemical Identifiers
| 7722-84-1 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| peroxol |
| H2O2 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |
Specifications
| Colorless | |
| 2.0,98.0,0.02 | |
| H2O2 | |
| OO | |
| 34.01 g/mol | |
| Reagent | |
| Stabilized | |
| 500 mL | |
| 3% (w/w) |
| 7722-84-1,7732-18-5,103-84-4 | |
| 4.0,98.0,0.02 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| peroxol | |
| 3% | |
| Amber Poly Bottle | |
| Aqueous Solution | |
| Liquid | |
| Hydrogen Peroxide |