missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, 3% (w/w), Stabilized Reagent Grade, Ricca Chemical

Supplier:  Ricca Chemical Company 381932

 View more versions of this product

Catalog No. 3819-32


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hydrogen Peroxide
7722-84-1,7732-18-5,103-84-4
4.0,98.0,0.02
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
OO
34.01
CHEBI:16240
Reagent
Stabilized
1 L
3% (w/w)
Colorless
2.0,96.0,0.02
H2O2
MHAJPDPJQMAIIY-UHFFFAOYSA-N
peroxol
784
3%
Amber Poly Bottle
Aqueous Solution
Liquid

Chemical Identifiers

7722-84-1
MHAJPDPJQMAIIY-UHFFFAOYSA-N
784
peroxol
H2O2
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
CHEBI:16240
OO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions