missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, 3% (w/w), Stabilized Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company 38195HP
Specifications
| Hydrogen Peroxide | |
| 7722-84-1,7732-18-5,103-84-4 | |
| 4.0,98.0,0.02 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| OO | |
| 34.01 | |
| CHEBI:16240 | |
| Reagent | |
| Stabilized | |
| 20 L | |
| 3% (w/w) |
| Colorless | |
| 2.0,96.0,0.02 | |
| H2O2 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| peroxol | |
| 784 | |
| 3% | |
| Hard Pack | |
| Aqueous Solution | |
| Liquid |
Chemical Identifiers
| 7722-84-1 | |
| MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
| 784 | |
| peroxol |
| H2O2 | |
| oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
| CHEBI:16240 | |
| OO |