missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide, 3% (w/w), Stabilized Reagent Grade, Ricca Chemical

Catalog No. 381916
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
55 gal.
Packaging:
Amber Poly Bottle
Black Poly Bottle
Hard Pack
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
381916 500 mL Amber Poly Bottle
381932 1 L Amber Poly Bottle
38191 4 L Black Poly Bottle
RIC01001SOL 20 L Hard Pack
381955 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 381916 Supplier Ricca Chemical Company Supplier No. 381916

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5 , 103-84-4
Molecular Formula H2O2
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
IUPAC Name peroxol
SMILES OO

Specifications

Color Colorless
CAS Min % 2.0, 98.0, 0.02
CAS Max % 4.0, 98.0, 0.02
Molecular Weight (g/mol) 34.01 g/mol
Percent Purity 3%
Grade Reagent
Packaging Amber Poly Bottle
Treatment(s) Stabilized
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 3% (w/w)
Chemical Name or Material Hydrogen Peroxide
Show More Show Less