missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.0200 N (N/50) in Isopropyl Alcohol, Ricca Chemical
Chemical Identifiers
| CAS | 7647-01-0 , 7732-18-5 , 7647-01-0 |
|---|---|
| Molecular Formula | ClH |
| Molecular Weight (g/mol) | 36.46 |
| MDL Number | MFCD00011324 |
| InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
| Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
| IUPAC Name | hydrogen chloride |
| SMILES | [H+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
R37920001A
|
N/A | 1 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
R379200020F
|
N/A | 20 L | Cubitainer |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| ClH | |
| MFCD00011324 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| [H+].[Cl-] |
Specifications
| 7647-01-0 , 7732-18-5 , 7647-01-0 | |
| 99.0, 2.36, 0.23 | |
| ClH | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| [H+].[Cl-] | |
| 36.46 |
| 96.0, 2.27, 0.22 | |
| Isopropyl Alcohol | |
| MFCD00011324 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| Hydrochloric Acid |