missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.0200 N (N/50) in Isopropyl Alcohol, Ricca Chemical
Supplier: Ricca Chemical Company R37920001A
| Quantity | 1 L |
|---|---|
| Packaging | Natural Poly Bottle |
Chemical Identifiers
| ClH | |
| MFCD00011324 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| [H+].[Cl-] |
Specifications
| Hydrochloric Acid | |
| 96.0, 2.27, 0.22 | |
| Natural Poly Bottle | |
| 1 L | |
| MFCD00011324 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride |