missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.0200 N (N/50) in Isopropyl Alcohol, Ricca Chemical

Supplier:  Ricca Chemical Company R379200020F

Quantity 20 L
Packaging Cubitainer

 View more versions of this product

Catalog No. R379200020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7647-01-0 , 7732-18-5 , 7732-18-5
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313  
hydrogen chloride
ClH
MFCD00011324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
CHEBI:23116
[H+].[Cl-]

Specifications

Hydrochloric Acid
0.22, 96.0, 2.27
Cubitainer
20 L
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313  
7647-01-0 , 7732-18-5 , 7732-18-5
0.23, 99.0, 2.36
Isopropyl Alcohol
ClH
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
[H+].[Cl-]
36.46
CHEBI:23116
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions