missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S.)
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
$309.06 - $1,351.48
Specifications
| Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
|---|---|
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
| Product Type | Fetal Bovine Serum |
| Sterility | Sterile |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity & Availability | ||||
|
SH3007102
|
Cytiva
SH30071.02 |
100 mL | None |
Each for $309.06
|
|
||||
|
SH3007103
|
Cytiva
SH30071.03 |
500 mL | None |
Each for $1,286.48
|
|
||||
|
SH3007103HI
|
Cytiva
SH30071.03HI |
500 mL | Heat Inactivated |
Each for $1,351.48
|
|
||||
|
SH300710340
|
Cytiva
SH30071.03IR2540 |
500 mL | Irradiated (25 to 40 doses) |
Each for $1,265.42
|
|
||||
Description
- Enhance growth of cells
- Tested for endotoxin, hemoglobin, and additional biochemical assays
- Filtered through serial 100 nm pore-size rated filters
Specifications
| Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
| Fetal Bovine Serum | |
| Sequential Triple-filtered, 100 nm | |
| ≤ 25 EU/mL | |
| HyClone | |
| USDA Inspected |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
| Sterile | |
| Characterized | |
| ≤ 25 mg/dL | |
| No |