missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S.)
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
Supplier: Cytiva SH30071.03IR2540
Description
- Enhance growth of cells
- Tested for endotoxin, hemoglobin, and additional biochemical assays
- Filtered through serial 100 nm pore-size rated filters
Chemical Identifiers
| Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
| Fetal Bovine Serum | |
| Sequential Triple-filtered, 100 nm | |
| ≤ 25 EU/mL | |
| HyClone | |
| USDA Inspected |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
| Sterile | |
| Characterized | |
| ≤ 25 mg/dL | |
| No |
Specifications
| Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride | |
| Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
| Fetal Bovine Serum | |
| Sterile | |
| Characterized | |
| Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
| No | |
| HyClone | |
| USDA Inspected |
| Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
| United States | |
| 500 mL | |
| Sequential Triple-filtered, 100 nm | |
| Irradiated (25 to 40 doses) | |
| ≤ 25 EU/mL | |
| ≤ 25 mg/dL | |
| No |