missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cell Boost 5™ Supplement

Catalog No. SH3086501
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3086501 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SH3086501 Supplier Cytiva Supplier No. SH30865.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides cell culture nutrients such as lipids, amino acids, vitamins, and growth factors. HyClone™ Cell Boost™ 5 Supplement is designed through Metabolic Pathway Design approach to support demand in the area of fed-batch cell culture and media customization.

  • Cell Boost 5 (CN-F) powdered supplement without L-glutamine
  • Non-sterile
  • Chemically defined
  • Free of animal-derived components
  • Tested in a variety of cell lines, including CHO, Hybridoma, NS0, and HEK 293

Specifications

Content And Storage Store at 2° to 8°C.
Product Type Cell Boost Supplement
Sterility Non-sterile
For Use With (Application) Bioprocessing, fed-batch cell culture, and media customization applications
Quantity 100 g